missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX5A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555646
This item is not returnable.
View return policy
Description
Immunogen sequence: QSVRCYSHGS QETDEEFDAR WVTYFNKPDI DAWELRKGIN TLVTYDMVPE PKIIDAALRA CRRLNDFAST VRILEVVKDK AGPHKEIYPY VIQELRPTLN ELGI Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The mitochondrially-encoded subunits perform the electron transfer of proton pumping activities. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. This gene encodes the nuclear-encoded subunit Va of the human mitochondrial respiratory chain enzyme. A pseudogene COX5AP1 has been found in chromosome 14q22.
Specifications
| COX5A | |
| Polyclonal | |
| Unconjugated | |
| COX5A | |
| AA959768; CcOX; COX; Cox5a; COX5A protein; COX-VA; cytochrome c oxidase polypeptide Va; cytochrome c oxidase polypeptide, mitochondrial; cytochrome c oxidase subunit 5A; cytochrome c oxidase subunit 5A, mitochondrial; cytochrome c oxidase subunit Va; cytochrome c oxidase, subunit Va; mitochondrial cytochrome c oxidase subunit 5a; mitochondrial cytochrome c oxidase subunit Va; VA | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9377 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P20674 | |
| COX5A | |
| Recombinant protein corresponding to Human COX5A. Recombinant protein control fragment (Product #RP-102244). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction