missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Polyclonal Antibody

COX4NB Polyclonal Antibody
Brand: Invitrogen PA580580
This item is not returnable.
View return policy
Description
COX4NB Polyclonal Antibody for Western Blot, IHC (P), IP
Specifications
| COX4NB | |
| Polyclonal | |
| Unconjugated | |
| Emc8 | |
| C16orf2; C16orf4; COX4 neighbor; COX4AL; COX4NB; Emc8; ER membrane protein complex subunit 8; FAM158B; family with sequence similarity 158, member B; neighbor of COX4; NOC4; Protein FAM158B | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 10328 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| O43402 | |
| Emc8 | |
| Recombinant Human EMC8/COX4NB/NOC4 protein (Met1-Cys210). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction