missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4I2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11463-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody was generated against full length COX4I2 protein.
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. COX4 (COX4I1) is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.Specifications
| COX4I2 | |
| Polyclonal | |
| Unconjugated | |
| COX4I2 | |
| COX IV 2, COX4, COX4 2, COX4B, COX4I2, COX4L2, COXIV 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84682, 84683, 84701 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q91W29, Q91Y94, Q96KJ9 | |
| Cox4i2 | |
| COX4I2 Fusion Protein Ag1987 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction