Learn More
COX4 Monoclonal Antibody (IB52C31H10), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£104.00 - £355.00
Specifications
Antigen | COX4 |
---|---|
Clone | IB52C31H10 |
Classification | Monoclonal |
Conjugate | Unconjugated |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15537676
|
Affymetrix eBioscience
14-9775-82 |
100 μg |
£355.00
100µg |
Estimated Shipment: 05-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15527676
|
Affymetrix eBioscience
14-9775-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Cytochrome c oxidase is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The mitochondrially-encoded subunits perform the electron transfer and proton pumping activities. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. This gene encodes the nuclear-encoded subunit IV isoform 1 of the human mitochondrial respiratory chain enzyme. It is located at the 3' of the NOC4 gene in a head-to-head orientation, and shares a promoter with it.
Cytochrome c oxidase (COX) is the terminal enzyme of the mitochondrial respiratory chain. It is a multi-subunit enzyme complex that couples the transfer of electrons from cytochrome c to molecular oxygen and contributes to a proton electrochemical gradient across the inner mitochondrial membrane. The complex consists of 13 mitochondrial- and nuclear-encoded subunits. The functions of the nuclear-encoded subunits are unknown but they may play a role in the regulation and assembly of the complex. COX4 (COX4I1) is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.Specifications
COX4 | |
Monoclonal | |
Liquid | |
RUO | |
COX4I1 | |
Primary | |
Antibody |
IB52C31H10 | |
Unconjugated | |
Mouse | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG1 κ | |
Affinity chromatography | |
COX4I1 |
For Research Use Only.