missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4 Antibody (CL3497) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-78718AF350
This item is not returnable.
View return policy
Description
COX4 Monoclonal antibody specifically detects COX4 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| COX4 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| COX IV-1, COX4-1, COX4COXIV, Cytochrome c oxidase polypeptide IV, cytochrome c oxidase subunit 4 isoform 1, mitochondrial, cytochrome c oxidase subunit IV isoform 1cytochrome c oxidase subunit IV, MGC72016 | |
| This antibody was developed using a recombinant protein derived from P13073, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Loading Controls, Neuroendocrinology | |
| 1327 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL3497 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction