missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ COX3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597297
This item is not returnable.
View return policy
Description
Immunogen sequence: MTHQTHAYHM VNPSPWPLTG AFSALLLTSG LVMWFHYNSI TLLTLGLLTN ILTMYQWWRD VIREGTYQGH HTPIVQKGLR YGMILFIVSE VFFFAGFFWA; Positive Samples: Mouse liver, Rat liver; Cellular Location: Mitochondrion inner membrane, Multi-pass membrane protein.
COX3 is a multi-pass membrane protein. It belongs to the cytochrome c oxidase subunit 3 family. Defects in COX3 are a cause of Leber hereditary optic neuropathy (LHON) and cytochrome c oxidase deficiency (COX deficiency). Defects in MT-CO3 are also found in mitochondrial encephalomyopathy with lactic acidosis and stroke-like episodes (MELAS) syndrome and recurrent myoglobinuria.
Specifications
| COX3 | |
| Polyclonal | |
| Unconjugated | |
| MT-CO3 | |
| ACI60_gp01; Co3; Coiii; COX3; COXIII; cytchrome c oxidase sububnit 3; Cytochrome c oxidase polypeptide III; Cytochrome c oxidase subunit 3; cytochrome c oxidase subunit III; mitochondrially encoded cytochrome c oxidase III; Mtco3; mt-Co3; OXI2; Q0275 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17710, 26204 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.36 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P00416, P05505 | |
| MT-CO3 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse MT-CO3 (NP_9043341). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction