missing translation for 'onlineSavingsMsg'
Learn More

COX17 Antibody (1A9), Novus Biologicals™

Product Code. 18356439 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18356439 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18356439 Supplier Novus Biologicals Supplier No. H00010063M03

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COX17 Monoclonal antibody specifically detects COX17 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX17
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 1A9
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005685
Gene Alias COX17 (yeast) homolog, cytochrome c oxidase assembly protein, COX17 cytochrome c oxidase assembly homolog (S. cerevisiae), cytochrome c oxidase assembly protein (S. cerevisiae), cytochrome c oxidase assembly protein (yeast), cytochrome c oxidase copper chaperone, human homolog of yeast mitochondrial copper recruitment, MGC104397, MGC117386
Host Species Mouse
Immunogen COX17 (NP_005685, 1 a.a. ~ 63 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MPGLVDSNPAPPESQEKKPLKPCCACPETKKARDACIIEKGEEHCGHLIEAHKECMRALGFKI
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 10063
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.