missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX16 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 19425-1-AP-150UL
This item is not returnable.
View return policy
Description
This protein is encoded by COX16, corresponding to the previously uncharacterized open reading frame YJL003w of the yeast genome. COX16 was identified in studies of COX-deficient mutants previously assigned to complementation group G22 of a collection of yeast pet mutants. To determine its location, Cox16p was tagged with a Myc epitope at the C terminus.Specifications
| COX16 | |
| Polyclonal | |
| Unconjugated | |
| COX16 | |
| C14orf112, COX16, HSPC203, PTD019 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51241, 66272 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.16 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9CR63, Q9P0S2 | |
| COX16 | |
| COX16 Fusion Protein Ag13756 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction