missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59560
This item is not returnable.
View return policy
Description
COX15 Polyclonal specifically detects COX15 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| COX15 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| COX15 (yeast) homolog, cytochrome c oxidase assembly protein, COX15 homolog, cytochrome c oxidase assembly protein (yeast), cytochrome c oxidase assembly protein COX15 homolog, cytochrome c oxidase subunit 15, EC 1.4.4.2, EC 3.1.3.5 | |
| Rabbit | |
| 44 kDa | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Yeast: 82%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast, Zebrafish | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1 mg/ml | |
| Western Blot 1.0 ug/ml, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
| Q7KZN9-2 | |
| COX15 | |
| Synthetic peptides corresponding to COX15(COX15 homolog, cytochrome c oxidase assembly protein (yeast)) The peptide sequence was selected from the N terminal of COX15. Peptide sequence DWHLIKEMKPPTSQEEWEAEFQRYQQFPEFKILNHDMTLTEFKFIWYMEY. | |
| Protein A purified | |
| RUO | |
| 1355 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction