missing translation for 'onlineSavingsMsg'
Learn More

COX15 Antibody (2D2), Novus Biologicals™

Product Code. 18338277 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338277 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18338277 Supplier Novus Biologicals Supplier No. H00001355M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

COX15 Monoclonal antibody specifically detects COX15 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen COX15
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 2D2
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_510870
Gene Alias COX15 (yeast) homolog, cytochrome c oxidase assembly protein, COX15 homolog, cytochrome c oxidase assembly protein (yeast), cytochrome c oxidase assembly protein COX15 homolog, cytochrome c oxidase subunit 15, EC 1.4.4.2, EC 3.1.3.5
Host Species Mouse
Immunogen COX15 (NP_510870, 92 a.a. ~ 152 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LTESGLSMVDWHLIKEMKPPTSQEEWEAEFQRYQQFPEFKILNHDMTLTEFKFIWYMEYSH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1355
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.