missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX11 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97240G
This item is not returnable.
View return policy
Description
COX11 Polyclonal antibody specifically detects COX11 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| COX11 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| COX11 (yeast) homolog, cytochrome c oxidase assembly protein, COX11 cytochrome c oxidase assembly homolog (yeast), COX11 homolog, cytochrome c oxidase assembly protein, COX11P, cytochrome c oxidase assembly protein COX11, mitochondrial, cytochrome c oxidase subunit 11 | |
| Produced in rabbits immunized with E. coli-derived Human COX11 fragment. | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 1353 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction