missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX-1 Rabbit anti-Human, Mouse, Rat, Clone: 8L4X10, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | COX-1 |
|---|---|
| Clone | 8L4X10 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18392235
|
Novus Biologicals
NBP3-16214-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18391165
|
Novus Biologicals
NBP3-16214-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COX-1 Monoclonal antibody specifically detects COX-1 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| COX-1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Immunology, Innate Immunity | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 5742 | |
| IgG | |
| Affinity purified |
| 8L4X10 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human COX-1 (P23219). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title