missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX-1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05599-20ul
This item is not returnable.
View return policy
Description
COX-1 Polyclonal antibody specifically detects COX-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| COX-1 | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS | |
| A synthetic peptide corresponding to a sequence within amino acids 415-514 of mouse COX-1 (NP_904330.1). AIMFVGVNMTFFPQHFLGLSGMPRRYSDYPDAYTTWNTVSSMGSFISLTAVLIMIFMIWEAFASKREVMSVSYASTNLEWLHGCPPPYHTFEEPTYVKVK | |
| 20 μg | |
| Cancer, Hypoxia, Immunology, Innate Immunity | |
| 5742 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction