missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX-1 Antibody (6B6), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | COX-1 |
|---|---|
| Clone | 6B6 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30036671
|
Novus Biologicals
NBP3-26362-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30028557
|
Novus Biologicals
NBP3-26362-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
COX-1 Monoclonal antibody specifically detects COX-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISASpecifications
| COX-1 | |
| Western Blot 1:500-1:5000, Flow Cytometry 1:20-1:200, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS | |
| A synthesized peptide derived from Human COX-1 [UniProt P23219] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 6B6 | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Rabbit | |
| Cancer, Hypoxia, Immunology, Innate Immunity | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 5742 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title