missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX-1 Antibody (345305), HRP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB3740H
408.85 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
COX-1 Monoclonal antibody specifically detects COX-1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| COX-1 | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 345305 | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| COX-1, COX1PGG/HS, COX3, Cyclooxygenase-1, EC 1.14.99.1, PCOX1, PGH synthase 1, PGHS1, PGHS-1PHS1, PHS 1, prostaglandin G/H synthase 1, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 1, Prostaglandin-endoperoxide synthase 1, prostaglandin-endoperoxide synthase 1 (prostaglandin G/H synthase andcyclooxygenase), PTGHS | |
| E. coli-derived recombinant human COX-1, Ala24-Leu599, Accession # P23219 | |
| 0.1 mL | |
| Cancer, Hypoxia, Immunology, Innate Immunity | |
| 5742 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction