missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Helios Italquartz Covers with Long Handle, Transparent
Click to view available options
For Use With (Equipment):
CR 10 Crucible
CR 13 Crucible
CR 14 Crucible
CR 16 Crucible
CR 2 Crucible
CR 6-7 Crucible
Unit Size:
Each
Description
- Transparent
- Include long handle
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Specifications
| Color | Transparent |
| Material | Quartz |
| Type | Covers with Long Handle |
| For Use With (Equipment) | CR 2 Crucible |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction