missing translation for 'onlineSavingsMsg'
Learn More

Helios Italquartz Covers with Long Handle, Translucent

Product Code. 15888064
Click to view available options
For Use With (Equipment):
CR 10 Crucible
CR 12 Crucible
CR 13 Crucible
CR 14 Crucible
CR 16 Crucible
CR 2 Crucible
CR 4 Crucible
CR 5 Crucible
CR 6-7 Crucible
CR 8 Crucible
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15888064

Brand: Helios Italquartz 64F00602

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Covers with long handle constructed of translucent, durable, resistant, and chemically inert quartz glass.

  • Translucent
  • Include long handle
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass

Specifications

Color Translucent
Material Quartz
Type Covers with Long Handle
For Use With (Equipment) CR 2 Crucible
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.