missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Coverglass Staining Racks

Product Code. 12627706 Shop All Epredia Products
Change view
Click to view available options
Description:
30-coverslip rack
Five-coverslip rack
Positions:
5
30
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Positions Unit Size
12627706 30-coverslip rack 30 Each
12617706 Five-coverslip rack 5 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12627706 Supplier Epredia™ Supplier No. 114
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Hold 22mm or 24mm coverslips securely with Epredia™ Coverglass Staining Racks. Available in two sizes.

  • Made of welded stainless steel

Five-coverslip Rack:

  • Fits in Vertical Staining Jar or Glass Beaker

Thirty-coverslip Rack:

  • Includes a spring-type detachable handle
  • Measures 4 x 11 x 1.2 in. (10.2 x 2.8 x 3.0 cm)
TRUSTED_SUSTAINABILITY

Specifications

Description 30-coverslip rack
Holds Slides
Material Stainless Steel
Positions 30
Length (Metric) Slide 22 or 24 mm
Product Type Coverslip Staining Rack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.