Learn More
KARTELL Cover for Microtitre Plates
Product Code. 12860923
Change view
Click to view available options
Plate Compatibility:
Plate Codes 2620, 2621, 2622
Unit Size:
Pack of 50
Product Code. Plate Compatibility unitSize
12860923 Plate Codes 2620, 2621, 2622 Pack of 50
1 options
Product Code. 12860923 Supplier KARTELL Supplier No. 02623/S
These Microtitre Plate Covers are compatible with flat-bottom, U-bottom and V-bottom well plates.
  • Sterile PS covers for Kartell Microtitrations plates 0262000, 0262200 and 262100
  • Available also in non sterile version (code 0262300).
Specifications
Color Clear
Material Polystyrene
Sterility Sterile
Adhesive None
Plate Compatibility Plate Codes 2620, 2621, 2622
Fit for Purpose Sterile