missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COUP-TF I/NR2F1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15974
This item is not returnable.
View return policy
Description
COUP-TF I/NR2F1 Polyclonal antibody specifically detects COUP-TF I/NR2F1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| COUP-TF I/NR2F1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| COUP transcription factor 1, COUP-TF1, COUP-TFI, EAR-3COUP transcription factor I, EAR3COUP-TF I, ERBAL3V-erbA-related protein 3, Nuclear receptor subfamily 2 group F member 1, nuclear receptor subfamily 2, group F, member 1, SVP44, TCFCOUP1, TFCOUP1NR2F2, transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-ahomolog-like 3) | |
| Rabbit | |
| 46 kDa | |
| 0.1 mL | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (100%), Rat (99%), Xenopus laevis (98%), Bovine (100%), Xenopus tropicalis (99%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot, Immunoprecipitation | |
| 0.92mg/mL | |
| Western Blot 1:500-1:3000, Immunoprecipitation | |
| P10589 | |
| NR2F1 | |
| Recombinant protein encompassing a sequence within the center region of human COUP TF1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 7025 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction