Learn More
Hirschmann™ Counting Chambers
Description
- Double net ruling
- Including 2 cover glasses for hemacytometer
- CE acc. IVD 98/79 EC
Specifications
Specifications
| Product Type | Counting Chamber (Burker) |
| Description | Double Net Ruling, Clamps, Bright Lined |
| Quantity | 1 Each |
| Depth (Metric) | 0.1 mm |
| Certifications/Compliance | CE acc. IVD 98/79 EC |
| Includes | 2 cover glasses for haemacytometer |