Learn More
Hirschmann™ Counting Chambers
Product Code. 10683002
Click to view available options
Product Type:
Counting Chamber (Burker)
Counting Chamber (Fuch-Rosenthal)
Counting Chamber (Nageotte)
Counting Chamber (Neubauer)
Counting Chamber (Thoma)
Counting Chamber (Thoma-neu)
Description:
Double Net Ruling, Bright Lined
Double Net Ruling, Clamps
Double Net Ruling, Clamps, Bright Lined
Unit Size:
Each
Product Code. 10683002
Brand: Hirschmann™ 8110206
Only null left
Add to basket
Add to basket
Facilitates quick and easy determination of cell concentration in liquid samples. Hirschmann™ Counting Chambers are available in a variety of depths to fit a range of systems.
  • Double net ruling
  • Including 2 cover glasses for hemacytometer
  • CE acc. IVD 98/79 EC
Specifications
Product Type Counting Chamber (Thoma-neu)
Description Double Net Ruling, Clamps, Bright Lined
Quantity 1 Each
Depth (Metric) 0.1 mm
Certifications/Compliance CE acc. IVD 98/79 EC
Includes 2 cover glasses for haemacytometer