missing translation for 'onlineSavingsMsg'
Learn More

Coumestrol, MedChemExpress

Product Code. 16449288
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16449288 10 mg 10mg
16401718 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16449288 Supplier Medchem Express Supplier No. HYN2335
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Coumestrol, a phytoestrogen present in soybean products, exhibits activities against cancers, neurological disorders, and autoimmune diseases. It suppresses proliferation of ES2 cells with an IC50 of 50 μM.

Coumestrol, a phytoestrogen present in soybean products, exhibits activities against cancers, neurological disorders, and autoimmune diseases. It suppresses proliferation of ES2 cells with an IC50 of 50 μM.

Specifications

Chemical Name or Material Coumestrol
CAS 479-13-0
Color Light Brown
Purity Grade Notes Research
Molecular Formula C15H8O5
Quantity 10 mg
Solubility Information DMSO : 25 mg/mL (93.21 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C2=C(OC3=CC(O)=CC=C32)C4=CC=C(O)C=C4O1
Molecular Weight (g/mol) 268.22
Formula Weight 268.22
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.