missing translation for 'onlineSavingsMsg'
Learn More

Coumarin, TRC

Product Code. 30367274
Change view
Click to view available options
Quantity:
50 mg
1 g
50 g
Unit Size:
1g
50g
50mg
Product Code. Quantity unitSize
30367274 50 mg 50mg
30359216 50 g 50g
30357686 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 30367274

Brand: Toronto Research Chemicals TRCC75538050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-64-5
Formula Weight 146.0368
InChI Formula InChI=1S/C9H6O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-6H
IUPAC Name chromen-2-one
Molecular Formula C9 H6 O2
Molecular Weight (g/mol) 146.14
Percent Purity >95%
SMILES O=C1Oc2ccccc2C=C1
Synonym Coumarin (8CI), 1,2-Benzopyrone, 2-Chromenone, 2-Propenoic acid, 3-(2-hydroxyphenyl)-, δ-lactone, 2H-Benzopyran-2-one, 2H-Chromen-2-one, 5,6-Benzo-2-pyrone, Benzo-α-pyrone, Coumarinic anhydride, Lympedim, NSC 8774, Rattex, Tonka bean camphor, cis-o-Coumarinic acid lactone, o-Hydroxycinnamic acid lactone
Chemical Name or Material Coumarin
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.