missing translation for 'onlineSavingsMsg'
Learn More

Coumarin, MedChemExpress

Product Code. 16489126
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16489126 100 mg 100mg
16472386 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16489126 Supplier Medchem Express Supplier No. HYN0709
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Coumarin is the primary bioactive ingredient in Radix Glehniae, named Beishashen in China, which possesses many pharmacological activities, including anticancer, anti-inflammation and antivirus activities.

Coumarin is the primary bioactive ingredient in Radix Glehniae, named Beishashen in China, which possesses many pharmacological activities, including anticancer, anti-inflammation and antivirus activities.

Specifications

Chemical Name or Material Coumarin
Physical Form Solid
CAS 91-64-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H6O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 100 mg/mL (684.28 mM) ∣H2O : 4 mg/mL (27.37 mM; Need ultrasonic)
SMILES O=C1C=CC2=CC=CC=C2O1
Molecular Weight (g/mol) 146.14
Formula Weight 146.14
Percent Purity 99.83%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.