missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 6, 98%, pure

Product Code. 10177270 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10177270

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 38215-36-0 | C20H18N2O2S | 350.436 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38215-36-0
Molecular Formula C20H18N2O2S
Molecular Weight (g/mol) 350.436
MDL Number MFCD00041869
InChI Key VBVAVBCYMYWNOU-UHFFFAOYSA-N
Synonym 3-(2'-Benzothiazolyl)-7-diethylaminocoumarin
PubChem CID 100334
ChEBI CHEBI:51942
IUPAC Name 3-(1,3-benzothiazol-2-yl)-7-(diethylamino)chromen-2-one
SMILES CCN(CC)C1=CC2=C(C=C1)C=C(C(=O)O2)C3=NC4=CC=CC=C4S3

Specifications

Assay Percent Range 98%
Fluorescence Max 497nm (in methanol)
Infrared Spectrum Authentic
Lambda Max 458nm (in methanol)
Formula Weight 350.44
Grade Extra Pure
Molar Absorption Coefficient 40800L/(mole.cm) (lambda max.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
UV_Vis 97.5% min.
Color Orange
Quantity 500 mg
Physical Form Powder
Chemical Name or Material Coumarin 6, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Coumarin 6, laser grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.