missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 343, pure, laser grade

Product Code. 10015710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10015710 500 mg Glass bottle 500mg
10249093 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10015710 Supplier Thermo Scientific Acros Supplier No. 405685000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55804-65-4 | C16H15NO4 | 285.299 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55804-65-4
Molecular Formula C16H15NO4
Molecular Weight (g/mol) 285.299
MDL Number MFCD00051335
InChI Key KCDCNGXPPGQERR-UHFFFAOYSA-N
PubChem CID 108770
ChEBI CHEBI:51941
SMILES C1CC2=C3C(=C4C(=C2)C=C(C(=O)O4)C(=O)O)CCCN3C1

Spezifikation

Melting Point 240.0°C
Fluorescence Max 470nm (in basic ethanol)
Solubility Information Solubility in water: slightly soluble.
Infrared Spectrum Authentic
Lambda Max 409nm (in basic ethanol)
Formula Weight 285.29
Grade Pure
Molar Absorption Coefficient 19900L/(mole.cm) (lambda max.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
Color Orange
Quantity 500 mg
Physical Form Crystalline Powder
Chemical Name or Material Coumarin 343
Mehr anzeigen Weniger anzeigen

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.