missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 2, 99%, laser grade

Product Code. 10566972 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
This item is not returnable. View return policy

Product Code. 10566972

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 26078-25-1 | C13H15NO2 | 217.268 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 168.0°C to 170.0°C
CAS 26078-25-1
Assay Percent Range 99%
Molecular Formula C13H15NO2
MDL Number MFCD00006860
Synonym 4, 6-Dimethyl-7-ethylaminocoumarin
Solubility Information Solubility in water: slightly soluble in water.
InChI Key QZXAEJGHNXJTSE-UHFFFAOYSA-N
Infrared Spectrum Authentic
SMILES CCNC1=C(C=C2C(=CC(=O)OC2=C1)C)C
IUPAC Name 7-(ethylamino)-4,6-dimethylchromen-2-one
Molecular Weight (g/mol) 217.268
PubChem CID 96929
Lambda Max 366nm (in absolute ethanol)
Formula Weight 217.27
Percent Purity 99%
Grade Extra Pure
Molar Absorption Coefficient 20000L/(mole.cm) (at lambda min.)
Packaging Glass bottle
Suitability (for use as a laser dye) passes
UV_Vis 98.5% min.
Color Green/Yellow to Yellow
Quantity 500 mg
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Coumarin 2, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Coumarin 2, laser grade
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.