missing translation for 'onlineSavingsMsg'
Learn More

Coumarin 1, TRC

Product Code. 30350831
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350831 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350831 Supplier Toronto Research Chemicals Supplier No. TRCC755508250MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Coumarin 1
CAS 91-44-1
Molecular Formula C14H17NO2
Quantity 250 mg
Synonym 7-(Diethylamino)-4-methyl-2H-1-benzopyran-2-one, 4-Methyl-7-(diethylamino)coumarin, 7-(Diethylamino)-4-methyl-2H-chromen-2-one, 7-(Diethylamino)-4-methylcoumarin, 7D4MC, Aclarat 8678, Blancol WNS, Blancophor AW, Blancophor FFG, Blankophor SOL, Brightener SWN, C 47, C 47 (coumarin derivative), C.I. 551100, C.I. Fluorescent Brightener 140, C.I. Fluorescent Brightener 52, C.I. Fluorescent Brightening Agent 52, Calcofluor RW, Calcofluor White RW, Calcofluor White SD, Coumarin 460, Coumarin 47, DEMC, Diethylamino-4-methylcoumarin, Fluorescent Brightener 140, Fluorescent Brightener 52, HR 1, Hakkol P, Hiltamine Arctic White SOL, Kayaphor WN, Leucophor WS, Leukophor WS, MDAC, NF-MC, NSC 61830, Neo-Super HR 1, Optiblanc SPL 10, Rylux VPA, Rylux VPA-T, Tinopal SWN, Uvitex SWN, Uvitex WGS, Whitefluor B, Whitex WS, 7-(Diethylamino)-4-methyl-coumarin
InChI Formula InChI=1S/C14H17NO2/c1-4-15(5-2)11-6-7-12-10(3)8-14(16)17-13(12)9-11/h6-9H,4-5H2,1-3H3
SMILES COC(C=C1)=CC=C1N2C(SCC3=NC(C4=CC=C(C)C=C4)=NO3)=NN=C2C5=CC=NC=C5
IUPAC Name 7-(diethylamino)-4-methylchromen-2-one
Molecular Weight (g/mol) 231.29
Formula Weight 231.1259
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.