Learn More
Coulter™ Whole Blood Lysing Reagent Kit
Product Code. 15207148
Encompass
Click to view available options
Quantity:
1 Kit
Product Code. 15207148
Brand: Coulter™ 6602764
Includes
Immunolyse, Fixative, and Stabilizer solutions
For the preparation of whole blood, bone marrow aspirates, and other sample types for analysis
  • 100–test kit prepares samples for analysis by fluorescence microscopy or flow cytometry
  • 300–test kit prepares samples for flow cytometry only
For In Vitro Diagnostic Use
Specifications
Certifications/Compliance IVD, CE, FCM
Content And Storage Immuno-lyse 1 x 6mL vial, fixative 2 x 20mL vial, stabilizer 1 1 x 6mL vial
Includes Immunolyse, fixative, and stabilizer solutions
No. of Tests 100
Quantity 1 Kit
Product Type Whole Blood Lysing Reagents Kit