missing translation for 'onlineSavingsMsg'
Learn More

Coulter™ Cyto-Trol™ Control Cells

Product Code. 15184303
Encompass
Click to view available options
:
Cytotrol cells
This item is not returnable. View return policy

Product Code. 15184303

Brand: Coulter™ 6604248

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Assess the activity of monoclonal antibodies by flow cytometry

Specifications

Certifications/Compliance IVD, CE, FCM
Content And Storage 2° to 8°C
Format Lyophilized
Includes 5 vials of lyophilized preparation human lymphocytes, five 1mL vials of reconstitution buffer
No. of Tests 50
Stability 24 hrs.
Product Type Cyto-Trol Control Cells Kit
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.