missing translation for 'onlineSavingsMsg'
Learn More

Cottonseed Oil, MP Biomedicals

Product Code. 11425532 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 lb.
25 lb.
Unit Size:
1lb
25lb
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425532 25 lb. 25lb
11415532 1 lb. 1lb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11425532 Supplier MP Biomedicals Supplier No. 0290141925

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Saponification Number: 190-198; Iodine Number: 105-114

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8001-29-4

Specifications

Quantity 25 lb.
Chemical Name or Material Cottonseed Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.