missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cotton Rat IL-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF584-SP
This item is not returnable.
View return policy
Description
IL-4 Polyclonal specifically detects IL-4 in Cotton Rat samples. It is validated for Western Blot, Neutralization.
Specifications
| IL-4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B cell growth factor 1, BCDF, B-cell stimulatory factor 1, BCGF1, BCGF-1, binetrakin, BSF1, BSF-1, IL4, IL-4B_cell stimulatory factor 1, IL4E12, interleukin 4, interleukin-4, Lymphocyte stimulatory factor 1, MGC79402, Pitrakinra | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3565 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.075-0.3 ug/mL | |
| AAL18820 | |
| IL4 | |
| E. coli-derived recombinant cotton rat IL-4 Cys21-Phe147 Accession # AAL18820 | |
| 25 μg | |
| Primary | |
| Detects cotton rat IL-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant rat IL-4 is observed, 5% cross-reactivity with recombinant mouse IL-4 is observed, and less than 1% cross-reactivity with recombinant human IL-4 and recombinant porcine IL-4 is observed. | |
| Cotton Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction