missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cotton Rat CXCL1/2/3/GRO Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB1114
This item is not returnable.
View return policy
Description
CXCL1/2/3/GRO Monoclonal specifically detects CXCL1/2/3/GRO in Cotton Rat samples. It is validated for Western Blot, Neutralization.
Specifications
| CXCL1/2/3/GRO | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Neutralization 0.08-0.4 ug/mL | |
| AAL16934 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2919 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization | |
| 186413 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 | |
| E. coli-derived recombinant cotton rat GRO Ala28-Lys100 Accession # AAL16934 | |
| 500 μg | |
| Primary | |
| Detects cotton rat CXCL1/2/3/GRO in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 5-20% cross-reactivity with recombinant rat CXCL1 and recombinant human CXCL2 and no cross-reactivity with recombinant cotton rat CXCL10, recombinant human CXCL1, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12/SDF-1 alpha , rh12/SDF-1 beta , 13, 16, recombinant mouse CXCL1, 2, 4, 6, 7, 9, 10, 11, 12/SDF-1 alpha , 13, 15, 16, 17, recombinant rat (rr) CXCL2, rrCXCL3/CINC-2 alpha , rr3/CINC-2 beta , or recombinant porcine CXCL8. | |
| Cotton Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction