missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cotton Rat CD4 Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6676S-100UG
This item is not returnable.
View return policy
Description
CD4 Monoclonal specifically detects CD4 in Cotton Rat samples. It is validated for Flow Cytometry.
Specifications
| CD4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects cotton rat CD4 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant CD4 from human, mouse, rat, cat, or dog is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| 695542 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD4 | |
| Mouse myeloma cell line NS0-derived recombinant cotton rat CD4 Lys26-Pro393 | |
| RUO | |
| 920 | |
| Cotton Rat | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction