missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Aratherma Nomex™ Heavy Gloves

Product Code. 12675333
Change view
Click to view available options
Size:
9
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
12675333 9 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12675333 Supplier Honeywell Safety Products™ Supplier No. 223207009

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Handle sharp, hot objects safely with these comfortable, shock-absorbing gloves. Honeywell Safety Products™ Aratherma Nomex™ Heavy Gloves are knitted and seamless and come with a seamless, cotton interlock liner for comfort and sweat absorption.

  • For handling sharp parts in a dry and hot atmosphere.
  • Protection against thermal hazards up to 250°C.
  • Seamless, 100% Nomex™ glove for superior thermal resistance.
  • Lined with a seamless cotton interlock glove that adds to mechanical resistance and absorbs perspiration.
  • Knitted for hand conformity, snugness, comfort, and dexterity.
  • The glove's relative thickness (gauge 13) enables it to absorb shocks, making it suitable for the handling of heavy objects.
  • Ambidextrous.
  • White color enhances hygiene and cleanliness.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical Hazards, EN 407-04 Thermical hazards, EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number 0075/797/162/12/07/0435, EC Attestation Number 0075/797/162/12/07/0435
Classification Dexterity: 2
Description Gauge 13
Color White
Cut Resistance Rating 4
Hand Ambidextrous
Material Nomex, Cotton
Product Type Gloves
Size 9
For Use With (Application) Protection against thermal hazards up to 250°C. For handling in a dry and hot atmosphere of sharp parts. Automotive, Glass, Iron and Steel industries.
Includes Oversewed knitted wrist
Liner Material Cotton
Lining Lined
Model T09
Quantity 10Pair/PK
Resistant To Heat
Tear Rating 4
Length (Metric) 26.5 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.