Learn More
COTL1 Mouse anti-Human, CoraLite 488, Clone: 4A3D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60237-100UL
Description
The COTL1 antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human COTL1. This antibody recognizes human, mouse, pig antigen. The COTL1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| COTL1 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 23406 | |
| IgG1 |
| Immunofluorescence | |
| 4A3D9 | |
| Mouse | |
| Primary | |
| Human, Pig, Mouse |