missing translation for 'onlineSavingsMsg'
Learn More

Cotinine, MedChemExpress

Product Code. 16444567
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444567 50 mg 50mg
16408666 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16444567 Supplier Medchem Express Supplier No. HYB1178
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cotinine ((-)-Cotinine), an alkaloid in tobacco and a major metabolite of nicotine, is used as a biological indicator to measure the composition of tobacco smoke

Cotinine ((-)-Cotinine), an alkaloid in tobacco and a major metabolite of nicotine, is used as a biological indicator to measure the composition of tobacco smoke

Specifications

Chemical Name or Material Cotinine
CAS 486-56-6
Color Light Brown
Purity Grade Notes Research
Molecular Formula C10H12N2O
Quantity 50 mg
Synonym (-)-Cotinine (S)-Cotinine NIH-10498
Solubility Information Ethanol : 120 mg/mL (680.97 mM; Need ultrasonic) ∣DMSO : 65 mg/mL (368.86 mM; Need ultrasonic)
SMILES O=C1N(C)[C@H](C2=CC=CN=C2)CC1
Molecular Weight (g/mol) 176.22
Formula Weight 176.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.