missing translation for 'onlineSavingsMsg'
Learn More

(-)-Cotinine 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30459953
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30459953 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30459953 Supplier LoGiCal Supplier No. LGCAMP0517.0301

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cotinine
CAS 486-56-6
Analyte or Component Names Cotinine
Shipping Condition Room Temperature
Molecular Formula C10 H12 N2 O
Certifications/Compliance ISO 17034
Synonym (5S)-1-Methyl-5-(pyridin-3-yl)pyrrolidin-2-one, Nicotine Imp. C (EP), Cotinine, (-)-Cotinine, Nicotine USP Related Compound C, Nicotine USP RC C, Nicotine Related Compound C, Nicotine Impurity C
InChI Formula InChI=1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/t9-/m0/s1
Solution Type Single Solution
SMILES CN1[C@@H](CCC1=O)c2cccnc2
IUPAC Name (5S)-1-methyl-5-pyridin-3-ylpyrrolidin-2-one
Solvent or Matrix Methanol
Industry Type Clinical Testing
Molecular Weight (g/mol) 176.22
Quantity 1 mL
Formula Weight 176.095 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.