missing translation for 'onlineSavingsMsg'
Learn More

COTI-2, MedChemExpress

Product Code. 16473706
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16473706 25 mg 25mg
16460418 5 mg 5mg
16457746 100 mg 100mg
16453887 50 mg 50mg
16427958 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16473706 Supplier Medchem Express Supplier No. HY19896

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress COTI-2, an anti-cancer drug with low toxicity, is an orally available third generation activator of p53 mutant forms. COTI-2 acts both by reactivating mutant p53 and inhibiting the PI3K/AKT/mTOR pathway. COTI-2 induces apoptosis in multiple human tumor cell lines. COTI-2 exhibits antitumor activity in HNSCC through p53-dependent and -independent mechanisms. COTI-2 converts mutant p53 to wild-type conformation.

COTI-2, an anti-cancer drug with low toxicity, is an orally available third generation activator of p53 mutant forms. COTI-2 acts both by reactivating mutant p53 and inhibiting the PI3K/AKT/mTOR pathway. COTI-2 induces apoptosis in multiple human tumor cell lines. COTI-2 exhibits antitumor activity in HNSCC through p53-dependent and -independent mechanisms. COTI-2 converts mutant p53 to wild-type conformation.

Specifications

Chemical Name or Material COTI-2
CAS 1039455-84-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C19H22N6S
Quantity 25 mg
Solubility Information DMSO : 5 mg/mL (13.64 mM; Need ultrasonic)
SMILES S=C(N1CCN(C2=NC=CC=C2)CC1)N/N=C3CCCC4=C\3N=CC=C4
Molecular Weight (g/mol) 366.48
Formula Weight 366.48
Percent Purity 98.48%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.