missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COTI-2, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
COTI-2, an anti-cancer drug with low toxicity, is an orally available third generation activator of p53 mutant forms. COTI-2 acts both by reactivating mutant p53 and inhibiting the PI3K/AKT/mTOR pathway. COTI-2 induces apoptosis in multiple human tumor cell lines. COTI-2 exhibits antitumor activity in HNSCC through p53-dependent and -independent mechanisms. COTI-2 converts mutant p53 to wild-type conformation.
Specifications
Specifications
| Chemical Name or Material | COTI-2 |
| CAS | 1039455-84-9 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C19H22N6S |
| Quantity | 50 mg |
| Solubility Information | DMSO : 5 mg/mL (13.64 mM; Need ultrasonic) |
| SMILES | S=C(N1CCN(C2=NC=CC=C2)CC1)N/N=C3CCCC4=C\3N=CC=C4 |
| Molecular Weight (g/mol) | 366.48 |
| Formula Weight | 366.48 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction