missing translation for 'onlineSavingsMsg'
Learn More

Costunolide, MedChemExpress

Product Code. 16411278
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16411278

Brand: Medchem Express HYN0036

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Costunolide ((+)-Costunolide) is a naturally occurring sesquiterpene lactone, with antioxidative, anti-inflammatory, antiallergic, bone remodeling, neuroprotective, hair growth promoting, anticancer, and antidiabetic properties. Costunolide can induce cell cycle arrest and apoptosis on breast cancer cells.

Costunolide ((+)-Costunolide) is a naturally occurring sesquiterpene lactone, with antioxidative, anti-inflammatory, antiallergic, bone remodeling, neuroprotective, hair growth promoting, anticancer, and antidiabetic properties. Costunolide can induce cell cycle arrest and apoptosis on breast cancer cells.

Specifications

Chemical Name or Material Costunolide
CAS 553-21-9
Color White
Purity Grade Notes Research
Molecular Formula C15H20O2
Quantity 5 mg
Synonym (+)-Costunolide Costus lactone
Solubility Information DMSO : ≥ 49 mg/mL (210.92 mM) ∣Ethanol : 25 mg/mL (107.61 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O[C@@]1([H])[C@@]2([H])CC/C(C)=C/CC/C(C)=C/1)C2=C
Molecular Weight (g/mol) 232.32
Formula Weight 232.32
Percent Purity 96.53%
Grade Research
Physical Form Crystals
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.