missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Costar™ Transwell™-COL Collagen-Coated PTFE Membrane Insert
GREENER_CHOICE

Product Code. 10136861 Shop All Corning Products
Change view
Click to view available options
Pore Size:
0.4 μm
3 μm
For Use With (Equipment):
12 Well Plate
24 Well Plate
6 Well Plate
Unit Size:
Pack of 24
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size For Use With (Equipment) unitSize
10136861 3 μm 6 Well Plate Pack of 24
10042501 0.4 μm 24 Well Plate Pack of 24
10457031 0.4 μm 12 Well Plate Pack of 24
10216581 0.4 μm 6 Well Plate Pack of 24
10005331 3 μm 12 Well Plate Pack of 24
10377791 3 μm 24 Well Plate Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10136861 Supplier Corning™ Supplier No. 3492

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Transparent, collagen-treated microporous membranes promote cell attachment and spreading

Permeable supports, also known as cell culture inserts, are an essential tool for the study of both anchorage-dependent and independent cell lines.

You can use cell culture inserts to:

  • Produce a cell culture environment that closely resembles an in vivo state
  • Allow polarized cells to carry out metabolic activities in a more natural manner because the cells feed both apically and basolaterally
  • Co-culture cells with or without cell-to-cell contact
  • Design a diversity of experiments using various pore sizes, membrane types, and coatings Transparent, Collagen-coated PTFE membrane
  • Promotes cell attachment and spreading
  • Equimolar mixture of Collagen type I and III
  • Individually packaged
  • Multiple well plates included in each case
  • Sterile

Order Info

Individually packed and supplied in package with tissue culture plates with lids; sterile.

TRUSTED_SUSTAINABILITY

Specifications

Culture Area 4.7 cm2
Material PTFE
Volume (Metric) Well 2.6 mL
Pore Size 3 μm
Type Membrane Insert
For Use With (Equipment) 6 Well Plate
Diameter (Metric) Filter 24 mm
No. per Pack 1/Pk.
No. per Case 24/Cs.
Surface Coating Collagen
Sterility Sterile
No. of Wells 6
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.