missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, U-Shaped-Bottom Microplate
GREENER_CHOICE

Product Code. 10420862 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10420862 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10420862 Supplier Corning™ Supplier No. 3360
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Clear; Sterility:Sterile; Lid:Without Lid

  • Medical-grade polystyrene
  • Well volume: 320μL to 360μL
  • Working volume: 75μL to 200μL
  • Choice of flat, round, flat Easy Wash™ or V-shaped bottom
  • Treated surface options are not coated, require no special handling and are stable at room temperature
  • Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
  • Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
  • Plate dimensions meet proposed industry standards

 

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Round Bottom (U-well)
For Use With (Application) Attachment and growth of anchorage-dependent cells
Quantity 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.