missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Costar™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate
Click to view available options
Packaging:
1/Pk
5/Pk
Description
- Cell culture microplates are sterile and nonpyrogenic.
- Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
Specifications
Format | Standard |
Lid | With Lid |
Culture Area | 0.32 cm2 |
Array | 8 x 12 |
Packaging | 5/Pk |
Binding Type | TC-Treated |
Color | Clear |
Material | Polystyrene |
No. of Wells | 96 |
Volume (Metric) Working | 0.1 to 0.2 mL |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'