missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well, Cell Culture-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE

Product Code. 10062511 Shop All Corning Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10062511 Clear Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10062511 Supplier Corning™ Supplier No. 3697
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Clear; Sterility:Sterile; Lid:With Lid

  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated

Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.

Cat. Nos. 3585 and 3595 have special low evaporation lids.

Cat. No. 3841 is aseptically manufactured.

TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Lid With Lid
Culture Area 0.16 cm2
Array 8 x 12
Packaging 20/Pk., 100/Cs.
Binding Type TC-Treated
Color Clear
Product Type Half Area
For Use With (Equipment) Plate readers
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 25 to 125 μL
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Enhance luminescent signals and have low background luminescence and fluorescence
No. per Case 100/Cs.
No. per Pack 20/Pk.
Quantity 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.