Learn More
Corning™ 96-Well Clear PVC Assay Microplates
Manufactured from clear flexible Polyvinyl Chloride (PVC) for economy and visibility. Corning™ 96-Well Clear PVC Assay Microplates present an excellent solution for laboratory miniaturization. Due to their flexible nature, these microplates are not compatible with automation.
Brand:  Corning™ 2595
Code : 66
Additional Details : Weight : 1.70640kg
 View more versions of this product
Product Code. 10691432
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Not treated PVC microplates are economical microplates for solution-based assays, serial dilutions, and general storage applications.
  • Well volume of 250 μL (260 μL for V-bottom); working well volume of 50 μL to 150 μL
  • Lids are not available.
  • Specifications
    Specifications
    Standard
    8 x 12
    PVC
    50 to 150 μL
    Non-treated
    Solution-based assays, serial dilutions and general storage applications
    25/Pk.
    Without Lid
    Clear
    96
    Non-sterile
    Flat Bottom (F-well)
    100/Cs.
    100/Cs.