Learn More
Corning™ Clear Polystyrene 96-Well EIA/RIA Microplates
GREENER_CHOICE
Product Code. 10380982 Shop All Corning Products
Change view
Click to view available options
Well Shape:
Flat Bottom (F-well)
Round
Binding Property:
High Binding
Medium Binding
Packaging:
25/Pk.
Individually Wrapped
Unit Size:
Pack of 100
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Well Shape Binding Property Packaging unitSize
10380982 Round Medium Binding 25/Pk. Pack of 100
10293532 Flat Bottom (F-well) High Binding 25/Pk. Pack of 100
10337891 Flat Bottom (F-well) Medium Binding Individually Wrapped Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10380982 Supplier Corning™ Supplier No. 3797
Only null left
Add to basket
Add to basket
Manufactured from clear medical grade polystyrene for strength and visibility. Corning™ Clear Polystyrene 96-Well Microplates present an excellent solution for cell culture and colorimetric (absorbance) assays. Use 96-well microplates for laboratory miniaturization and automation.
  • Cell culture microplates are sterile and nonpyrogenic.
  • Lids available where indicated
Cat. nos. 3798 and 3898 processed to improve hydrophilicity for hemagglutination and similar assays.
Cat. Nos. 3585 and 3595 have special low evaporation lids.
Cat. No. 3841 is aseptically manufactured.
TRUSTED_SUSTAINABILITY
Specifications
Binding Property Medium Binding
Format Standard
Lid Without Lid
Array 8 x 12
Packaging 25/Pk.
Binding Type Not Treated
Color Clear
Material Polystyrene
No. of Wells 96
Volume (Metric) Working 75 to 200 μL
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Round
For Use With (Application) Hydrophobic in nature and binds bio-molecules through passive interactions. Suitable for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions.
No. per Case 100/Cs.
No. per Pack 25/Pk.
Quantity 4 x 25