missing translation for 'onlineSavingsMsg'
Learn More

Corning™ 96-Well Solid Black or White Polystyrene Microplates
GREENER_CHOICE

Product Code. 10133811 Shop All Corning Products
Change view
Click to view available options
Color:
Black
White
Surface Treatment:
High Binding
Non-treated
Unit Size:
Pack of 100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Surface Treatment unitSize
10133811 Black High Binding Pack of 100
10679141 White High Binding Pack of 100
10002551 White Non-treated Pack of 100
10022561 Black Non-treated Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10133811 Supplier Corning™ Supplier No. 3925
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Manufactured from medical grade polystyrene for strength. Corning™ 96-Well Black or White Polystyrene Microplates are used for florescent and luminescent assays. Lot tested for opacity and low fluorescent/luminescent background.

  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Format Standard
    Lid Without Lid
    Array 8 x 12
    Packaging 4 inner packs of 25
    Color Black
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 75 to 200 μL
    Sterility Non-sterile
    Surface Treatment High Binding
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.