missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molded polystyrene with opaque walls and optically clear bottoms
- Bottoms are 60% thinner than conventional polystyrene microplates for lower background flourecence and enabling readings down to 340nm
- Opaque black or white walls prevent well to well crosstalk
- Recommended working volumes of 75 to 200μL
- Available untreated for solution-based assays, with high binding for immunoassay applications, or tissue culture treated for cell growth
- Treated surface options are not coated, require no special handling and are stable at room temperature
- Sterile microplates are sterilized by gamma radiation and certified nonpyrogenic
- Ideal for serial dilutions, hemagglutination or precipitation assays, and general storage applications
- Plate dimensions meet proposed industry standards
Order Info
Cat. No. 07-200-568 also available in a pack of 25.
Specifications
Specifications
| Format | Standard |
| Lid | Without Lid |
| Array | 8 x 12 |
| Color | Black, Clear Bottom |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 75 to 200 μL |
| Sterility | Non-sterile |
| Surface Treatment | High Binding |
| Well Shape | Flat Bottom (F-well) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
